Product Details
- SNP ID
-
rs200334193
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:122173943 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACGGACGGGCAACGTGTGGGAGGC[C/G]AGCCAGCCTCCCAGGCAGCAGAACA
- Phenotype
-
MIM: 609509
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IL31
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56719421] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL31
- Gene Name
- interleukin 31
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001014336.1 |
93 |
Silent Mutation |
CTC,CTG |
L22L |
NP_001014358.1 |
- Gene
- LRRC43
- Gene Name
- leucine rich repeat containing 43
View Full Product Details