Product Details
- SNP ID
-
rs199947250
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:55937494 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGACACCTAAGCCTGGCACTGTT[C/T]CAATCCTTTGAGACTGGTCACTGCT
- Phenotype
-
MIM: 125855
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DGKA
PubMed Links
Gene Details
- Gene
- DGKA
- Gene Name
- diacylglycerol kinase alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001345.4 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
NP_001336.2 |
NM_201444.2 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
NP_958852.1 |
NM_201445.1 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
NP_958853.1 |
NM_201554.1 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
NP_963848.1 |
XM_005268688.2 |
306 |
Silent Mutation |
TTC,TTT |
F113F |
XP_005268745.1 |
XM_005268689.2 |
306 |
UTR 5 |
|
|
XP_005268746.1 |
XM_005268690.2 |
306 |
UTR 5 |
|
|
XP_005268747.1 |
XM_011537991.2 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
XP_011536293.1 |
XM_011537993.2 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
XP_011536295.1 |
XM_011537995.2 |
306 |
UTR 5 |
|
|
XP_011536297.1 |
XM_017018900.1 |
306 |
Silent Mutation |
TTC,TTT |
F113F |
XP_016874389.1 |
XM_017018901.1 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
XP_016874390.1 |
XM_017018902.1 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
XP_016874391.1 |
XM_017018903.1 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
XP_016874392.1 |
XM_017018904.1 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
XP_016874393.1 |
XM_017018905.1 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
XP_016874394.1 |
XM_017018906.1 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
XP_016874395.1 |
XM_017018907.1 |
306 |
Silent Mutation |
TTC,TTT |
F75F |
XP_016874396.1 |
XM_017018908.1 |
306 |
Silent Mutation |
TTC,TTT |
F113F |
XP_016874397.1 |
XM_017018909.1 |
306 |
UTR 5 |
|
|
XP_016874398.1 |
- Gene
- PYM1
- Gene Name
- PYM homolog 1, exon junction complex associated factor
There are no transcripts associated with this gene.
View Full Product Details