Product Details

SNP ID
rs199770498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99516249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGGTCCTCCTCACAGCTGTCC[C/T]GGAGCTGCCGGTCCAGATTCTCGTG
Phenotype
MIM: 603544
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC85C PubMed Links

Gene Details

Gene
CCDC85C
Gene Name
coiled-coil domain containing 85C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144995.1 1540 Missense Mutation CAG,CGG Q370R NP_001138467.1
XM_011536705.2 1540 Missense Mutation CAG,CGG Q383R XP_011535007.1
XM_011536706.2 1540 Intron XP_011535008.1
XM_011536707.2 1540 Intron XP_011535009.1
Gene
CCNK
Gene Name
cyclin K
There are no transcripts associated with this gene.

View Full Product Details