Product Details

SNP ID
rs201138394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23042689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACTGCCCTCTGCGGCTGGGACC[A/G]CTCTGGCCCTGAGCTCTTCTACGTC
Phenotype
MIM: 611137 MIM: 600306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH24 PubMed Links

Gene Details

Gene
CDH24
Gene Name
cadherin 24
There are no transcripts associated with this gene.

Gene
PSMB11
Gene Name
proteasome subunit beta 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099780.1 523 Missense Mutation CAC,CGC H155R NP_001093250.1
Gene
PSMB5
Gene Name
proteasome subunit beta 5
There are no transcripts associated with this gene.

View Full Product Details