Product Details
- SNP ID
-
hCV190122919
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:78542623 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTCTTGATGAAGTCTTTTTTTCT[G/A]AAAAAATTTATAAACTCAATGAGTA
- Phenotype
-
MIM: 614681
MIM: 176846
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HYKK
PubMed Links
Gene Details
- Gene
- HYKK
- Gene Name
- hydroxylysine kinase
There are no transcripts associated with this gene.
- Gene
- PSMA4
- Gene Name
- proteasome subunit alpha 4
There are no transcripts associated with this gene.
View Full Product Details