Product Details

SNP ID
rs201796033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:98441140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGTCCTCGCATGACGGCTCAC[A/G]TCCTCGGAATTTCCTGGGTGTGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM169B PubMed Links

Gene Details

Gene
FAM169B
Gene Name
family with sequence similarity 169 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182562.2 640 Silent Mutation GAC,GAT D130D NP_872368.2

View Full Product Details