Product Details
- SNP ID
-
rs200149562
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43335110 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATCATTTTTTTCCTGCTAAAAT[G/T]ATAGAGGCAGAAGAGCAACAGCCTT
- Phenotype
-
MIM: 611246
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADAL
PubMed Links
Gene Details
- Gene
- ADAL
- Gene Name
- adenosine deaminase like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001012969.3 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
NP_001012987.1 |
NM_001159280.2 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
NP_001152752.1 |
NM_001324364.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
NP_001311293.1 |
NM_001324365.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
NP_001311294.1 |
NM_001324366.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
NP_001311295.1 |
NM_001324367.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
NP_001311296.1 |
NM_001324368.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
NP_001311297.1 |
XM_011521291.2 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
XP_011519593.1 |
XM_017021964.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
XP_016877453.1 |
XM_017021965.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
XP_016877454.1 |
XM_017021966.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
XP_016877455.1 |
XM_017021967.1 |
608 |
Intron |
|
|
XP_016877456.1 |
XM_017021968.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
XP_016877457.1 |
XM_017021969.1 |
608 |
Missense Mutation |
ATG,ATT |
M1I |
XP_016877458.1 |
- Gene
- LCMT2
- Gene Name
- leucine carboxyl methyltransferase 2
There are no transcripts associated with this gene.
View Full Product Details