Product Details

SNP ID
rs199651156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34348047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCCCTGCACTTCCCTTTCTCC[A/C]ACCAACTTCTGACCCACCAGACCAC
Phenotype
MIM: 606471 MIM: 608963 MIM: 604878
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NOP10 PubMed Links

Gene Details

Gene
NOP10
Gene Name
NOP10 ribonucleoprotein
There are no transcripts associated with this gene.

Gene
NUTM1
Gene Name
NUT midline carcinoma family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284292.1 561 Missense Mutation CAA,CCA Q60P NP_001271221.1
NM_001284293.1 561 Missense Mutation CAA,CCA Q50P NP_001271222.1
NM_175741.2 561 Missense Mutation CAA,CCA Q32P NP_786883.1
Gene
SLC12A6
Gene Name
solute carrier family 12 member 6
There are no transcripts associated with this gene.

View Full Product Details