Product Details

SNP ID
hCV190202812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1361762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGGACCCGTGCATCTCTTCCG[A/G]CTCTCGGGCAAGTGCTTCAGCCTGG
Phenotype
MIM: 607838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNPTG PubMed Links

Gene Details

Gene
GNPTG
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
There are no transcripts associated with this gene.

Gene
TSR3
Gene Name
TSR3, acp transferase ribosome maturation factor
There are no transcripts associated with this gene.

Gene
UNKL
Gene Name
unkempt family like zinc finger
There are no transcripts associated with this gene.

View Full Product Details