Product Details

SNP ID
rs200867929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84566932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCCAGCTCCTTCCGATCACT[G/T]ATCACAAACTCCTTAGCGAAATTCT
Phenotype
MIM: 606748
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COTL1 PubMed Links

Gene Details

Gene
COTL1
Gene Name
coactosin like F-actin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021149.3 524 Silent Mutation ATA,ATC I114I NP_066972.1

View Full Product Details