Product Details

SNP ID
rs199669467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:799038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCCAGCTGGTACTTGAGGCTCT[C/T]CACCTCTTTCTTCATCTGTGGCACG
Phenotype
MIM: 613201 MIM: 607298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHTF18 PubMed Links

Gene Details

Gene
CHTF18
Gene Name
chromosome transmission fidelity factor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022092.2 142 Intron NP_071375.1
XM_005255470.1 142 Intron XP_005255527.1
XM_005255471.3 142 Intron XP_005255528.1
XM_011522572.1 142 Intron XP_011520874.1
XM_011522573.1 142 Intron XP_011520875.1
XM_017023532.1 142 Intron XP_016879021.1
XM_017023533.1 142 Intron XP_016879022.1
XM_017023534.1 142 Intron XP_016879023.1
Gene
GNG13
Gene Name
G protein subunit gamma 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016541.2 142 Missense Mutation AAG,GAG K14E NP_057625.1
Gene
PRR25
Gene Name
proline rich 25
There are no transcripts associated with this gene.

View Full Product Details