Product Details

SNP ID
rs201538561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2237866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGCAGTGAGGTCTTCAAGTGG[C/T]TCATCCCTGACAGCGCCGACACCAC
Phenotype
MIM: 602622 MIM: 603022 MIM: 600305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNASE1L2 PubMed Links

Gene Details

Gene
DNASE1L2
Gene Name
deoxyribonuclease I-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301680.1 801 Missense Mutation CTC,TTC L231F NP_001288609.1
NM_001374.2 801 Missense Mutation CTC,TTC L231F NP_001365.1
XM_011522399.1 801 Missense Mutation CTC,TTC L322F XP_011520701.1
Gene
E4F1
Gene Name
E4F transcription factor 1
There are no transcripts associated with this gene.

Gene
ECI1
Gene Name
enoyl-CoA delta isomerase 1
There are no transcripts associated with this gene.

View Full Product Details