Product Details

SNP ID
hCV190270294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81899937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGTAGGCAAGGTGCCCGGCGAC[G/A]ACTGCCCGCTGGTGTGGGGCCAGTG
Phenotype
MIM: 604171 MIM: 614534 MIM: 607996 MIM: 602679
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ALYREF PubMed Links

Gene Details

Gene
ALYREF
Gene Name
Aly/REF export factor
There are no transcripts associated with this gene.

Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
There are no transcripts associated with this gene.

Gene
NPB
Gene Name
neuropeptide B
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
There are no transcripts associated with this gene.

View Full Product Details