Product Details

SNP ID
rs202129528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1511128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGCCAAGGGAAGCCGCCGCCGC[A/G]TCTCCTCCAGGAATCTGTAGGCCTG
Phenotype
MIM: 614620 MIM: 611140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT140 PubMed Links

Gene Details

Gene
IFT140
Gene Name
intraflagellar transport 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014714.3 3287 Missense Mutation ACG,ATG T1402M NP_055529.2
XM_005255725.4 3287 Intron XP_005255782.1
XM_005255726.3 3287 Intron XP_005255783.1
XM_006720990.3 3287 Missense Mutation ACG,ATG T1402M XP_006721053.1
XM_006720991.3 3287 Missense Mutation ACG,ATG T1402M XP_006721054.1
XM_006720992.3 3287 Missense Mutation ACG,ATG T613M XP_006721055.1
XM_011522766.2 3287 Missense Mutation ACG,ATG T1320M XP_011521068.1
XM_011522767.2 3287 Missense Mutation ACG,ATG T1077M XP_011521069.1
XM_011522769.2 3287 Intron XP_011521071.1
XM_011522771.2 3287 Intron XP_011521073.1
XM_011522772.2 3287 Intron XP_011521074.1
XM_017023910.1 3287 Missense Mutation ACG,ATG T1402M XP_016879399.1
XM_017023911.1 3287 Missense Mutation ACG,ATG T797M XP_016879400.1
Gene
TELO2
Gene Name
telomere maintenance 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016111.3 3287 Intron NP_057195.2
XM_011522773.2 3287 Intron XP_011521075.1
XM_011522774.2 3287 Intron XP_011521076.1
XM_011522775.2 3287 Intron XP_011521077.1
XM_011522776.2 3287 Intron XP_011521078.1
XM_011522777.2 3287 Intron XP_011521079.1
XM_011522778.2 3287 Intron XP_011521080.1
XM_017023914.1 3287 Intron XP_016879403.1

View Full Product Details