Product Details

SNP ID
rs201816896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTTGTCCATGCTGGGGCTGGT[A/G]AGGGTCATGTCATCGCTGCTGGCTG
Phenotype
MIM: 156565 MIM: 600928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPG PubMed Links

Gene Details

Gene
MPG
Gene Name
N-methylpurine DNA glycosylase
There are no transcripts associated with this gene.

Gene
NPRL3
Gene Name
NPR3 like, GATOR1 complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039476.2 1698 Silent Mutation CTC,CTT L279L NP_001034565.1
NM_001077350.2 1698 Silent Mutation CTC,CTT L458L NP_001070818.1
NM_001243247.1 1698 Silent Mutation CTC,CTT L380L NP_001230176.1
NM_001243248.1 1698 Silent Mutation CTC,CTT L433L NP_001230177.1
NM_001243249.1 1698 Silent Mutation CTC,CTT L433L NP_001230178.1

View Full Product Details