Product Details

SNP ID
rs199667738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75891061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCTGTCCTCCTGGACGCAGAG[C/T]CCCCAGGAGCAGGGTCCCCGGCCAA
Phenotype
MIM: 611844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL38 PubMed Links

Gene Details

Gene
MRPL38
Gene Name
mitochondrial ribosomal protein L38
There are no transcripts associated with this gene.

Gene
TRIM65
Gene Name
tripartite motif containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256124.1 1309 Silent Mutation NP_001243053.1
NM_173547.3 1309 Silent Mutation NP_775818.2
XM_006721760.3 1309 Silent Mutation XP_006721823.1
XM_006721761.3 1309 Silent Mutation XP_006721824.1
XM_011524499.2 1309 Intron XP_011522801.1
XM_011524500.2 1309 Intron XP_011522802.1
XM_011524501.2 1309 Intron XP_011522803.1
XM_011524502.2 1309 Intron XP_011522804.1
XM_011524503.2 1309 Intron XP_011522805.1
XM_017024343.1 1309 Intron XP_016879832.1
XM_017024344.1 1309 UTR 3 XP_016879833.1

View Full Product Details