Product Details

SNP ID
rs200071006
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81894621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCATGCTGTCTGAGCGGCCCC[A/G]ACTGTGAGTGTCCCCTCTGTGCTGT
Phenotype
MIM: 604171 MIM: 614534 MIM: 607996 MIM: 602679
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ALYREF PubMed Links
Additional Information
For this assay, SNP(s) [rs149073349] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ALYREF
Gene Name
Aly/REF export factor
There are no transcripts associated with this gene.

Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002244.2 276 Intron NP_001002244.1
NM_001002245.2 276 Intron NP_001002245.1
NM_001002246.2 276 Intron NP_001002246.1
NM_001002247.2 276 Intron NP_001002247.1
NM_001002248.2 276 Intron NP_001002248.1
NM_001002249.2 276 Intron NP_001002249.1
NM_001289414.1 276 Intron NP_001276343.1
NM_001289415.1 276 Intron NP_001276344.1
NM_001289416.1 276 Intron NP_001276345.1
NM_001289417.1 276 Intron NP_001276346.1
NM_001289418.1 276 Missense Mutation CAA,CGA Q29R NP_001276347.1
NM_001289419.1 276 Missense Mutation CAA,CGA Q29R NP_001276348.1
NM_001289420.1 276 Intron NP_001276349.1
NM_016476.11 276 Intron NP_057560.8
Gene
NPB
Gene Name
neuropeptide B
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
There are no transcripts associated with this gene.

View Full Product Details