Product Details

SNP ID
rs200991847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29607768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACATCGAGCAGCTGGATCCCCG[C/T]GGCCGGACTCCCCTGCACCTGGCCA
Phenotype
MIM: 615124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13B PubMed Links

Gene Details

Gene
ANKRD13B
Gene Name
ankyrin repeat domain 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152345.4 254 Silent Mutation CGC,CGT R47R NP_689558.4
XM_005257920.4 254 Silent Mutation CGC,CGT R47R XP_005257977.1
XM_011524319.2 254 Silent Mutation CGC,CGT R39R XP_011522621.1
XM_011524320.2 254 Silent Mutation CGC,CGT R47R XP_011522622.1
XM_011524321.2 254 Silent Mutation CGC,CGT R47R XP_011522623.1
XM_017024174.1 254 UTR 5 XP_016879663.1
XM_017024175.1 254 UTR 5 XP_016879664.1
XM_017024176.1 254 UTR 5 XP_016879665.1
Gene
CORO6
Gene Name
coronin 6
There are no transcripts associated with this gene.

View Full Product Details