Product Details

SNP ID
rs200686073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67044824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCGGCCCACTTCGGCCTCTGGC[A/G]GATTTGTACCAAGCGCATCCCCATG
Phenotype
MIM: 114209
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNG1 PubMed Links

Gene Details

Gene
CACNG1
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000727.3 289 Missense Mutation CAG,CGG Q55R NP_000718.1

View Full Product Details