Product Details
- SNP ID
-
rs200687206
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8073757 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGGTTCGCAAGCACCTAGGGAAG[C/T]CTGACCGGCGGGGGAAAAGCCCAGG
- Phenotype
-
MIM: 603741
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALOX12B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12937410] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALOX12B
- Gene Name
- arachidonate 12-lipoxygenase, 12R type
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001139.2 |
1916 |
Missense Mutation |
GAC,GGC |
D552G |
NP_001130.1 |
View Full Product Details