Product Details
- SNP ID
-
rs201969522
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.17:40794013 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAGCTGCGTACAGTAGTTGCTCT[C/T]GGTCTCTGTCAAGGAGCACTCCAGG
- Phenotype
-
MIM: 616677
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT28
PubMed Links
Gene Details
- Gene
- KRT28
- Gene Name
- keratin 28
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181535.3 |
1078 |
Missense Mutation |
AAG,GAG |
K338E |
NP_853513.2 |
View Full Product Details