Product Details

SNP ID
rs201801331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6425699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGCCATGCGGTTCTCCAGCAGCC[C/T]CAGCTCCCTGCGCACCGCCTTCTGC
Phenotype
MIM: 604392
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIPL1 PubMed Links

Gene Details

Gene
AIPL1
Gene Name
aryl hydrocarbon receptor interacting protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033054.2 939 Missense Mutation AGG,GGG R243G NP_001028226.1
NM_001033055.2 939 Missense Mutation AGG,GGG R246G NP_001028227.1
NM_001285399.2 939 Missense Mutation AGG,GGG R294G NP_001272328.1
NM_001285400.2 939 Missense Mutation AGG,GGG R284G NP_001272329.1
NM_001285401.2 939 Missense Mutation AGG,GGG R282G NP_001272330.1
NM_001285402.1 939 Missense Mutation AGG,GGG R267G NP_001272331.1
NM_001285403.2 939 Intron NP_001272332.1
NM_014336.4 939 Missense Mutation AGG,GGG R306G NP_055151.3

View Full Product Details