Product Details

SNP ID
rs202046182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67044821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGAGGCGGCCCACTTCGGCCTCT[G/T]GCGGATTTGTACCAAGCGCATCCCC
Phenotype
MIM: 114209
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CACNG1 PubMed Links

Gene Details

Gene
CACNG1
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000727.3 286 Missense Mutation TGG,TTG W54L NP_000718.1

View Full Product Details