Product Details

SNP ID
rs202175535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9887823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAATGGAGTTCCCGGAGGGGGA[A/C]AAGGTGAAAGTGATCCTGAGCAAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TXNDC2 PubMed Links

Gene Details

Gene
TXNDC2
Gene Name
thioredoxin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098529.1 1793 Missense Mutation GAA,GAC E448D NP_001091999.1
NM_032243.5 1793 Missense Mutation GAA,GAC E381D NP_115619.4
XM_017026041.1 1793 Missense Mutation GAA,GAC E381D XP_016881530.1

View Full Product Details