Product Details

SNP ID
rs199637969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40742541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCCGTCAGTGGCCCGTGAGGG[C/T]GAGAAGTCCGTCAGCTGCAGGTTGC
Phenotype
MIM: 606476 MIM: 182285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198476.3 971 Silent Mutation NP_940878.3
XM_005258776.4 971 UTR 3 XP_005258833.1
XM_005258778.4 971 Silent Mutation XP_005258835.1
XM_006723152.2 971 UTR 3 XP_006723215.1
XM_006723153.2 971 UTR 3 XP_006723216.1
XM_011526774.2 971 Silent Mutation XP_011525076.1
XM_011526775.1 971 Silent Mutation XP_011525077.1
XM_011526776.2 971 Intron XP_011525078.1
XM_011526777.2 971 Silent Mutation XP_011525079.1
XM_011526778.2 971 Intron XP_011525080.1
XM_011526779.2 971 Intron XP_011525081.1
XM_011526782.2 971 Intron XP_011525084.1
XM_011526783.2 971 Intron XP_011525085.1
XM_011526784.2 971 Silent Mutation XP_011525086.1
XM_011526785.2 971 Silent Mutation XP_011525087.1
XM_011526786.2 971 Intron XP_011525088.1
XM_017026620.1 971 Intron XP_016882109.1
XM_017026621.1 971 Silent Mutation XP_016882110.1
XM_017026622.1 971 Intron XP_016882111.1
XM_017026623.1 971 Intron XP_016882112.1
XM_017026624.1 971 Silent Mutation XP_016882113.1
XM_017026625.1 971 Intron XP_016882114.1
XM_017026626.1 971 Silent Mutation XP_016882115.1
XM_017026627.1 971 Silent Mutation XP_016882116.1
Gene
ITPKC
Gene Name
inositol-trisphosphate 3-kinase C
There are no transcripts associated with this gene.

Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
There are no transcripts associated with this gene.

View Full Product Details