Product Details

SNP ID
rs199608846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14473417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGATGCACGACACCACCATGA[A/T]ACCGACAAGCTGGCCCACCATCTCC
Phenotype
MIM: 605072 MIM: 601032 MIM: 176802
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GIPC1 PubMed Links

Gene Details

Gene
GIPC1
Gene Name
GIPC PDZ domain containing family member 1
There are no transcripts associated with this gene.

Gene
PKN1
Gene Name
protein kinase N1
There are no transcripts associated with this gene.

Gene
PTGER1
Gene Name
prostaglandin E receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000955.2 1022 Missense Mutation ATC,TTC I302F NP_000946.2

View Full Product Details