Product Details

SNP ID
rs199662090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36818741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGATTTCCCACATTCTTTACATA[C/T]GTAAGGTTCCTCACCAGTATGCATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF790 PubMed Links

Gene Details

Gene
ZNF790
Gene Name
zinc finger protein 790
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242800.1 1764 Missense Mutation ATA,GTA I535V NP_001229729.1
NM_001242801.1 1764 Missense Mutation ATA,GTA I535V NP_001229730.1
NM_001242802.1 1764 Missense Mutation ATA,GTA I535V NP_001229731.1
NM_206894.3 1764 Missense Mutation ATA,GTA I535V NP_996777.2
XM_005258903.4 1764 Missense Mutation ATA,GTA I535V XP_005258960.1
XM_011526950.2 1764 Missense Mutation ATA,GTA I535V XP_011525252.1
Gene
ZNF790-AS1
Gene Name
ZNF790 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details