Product Details
- SNP ID
-
rs200418605
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39539975 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCGGCCTCCTCAGGCTGTGCCGG[A/G]GCCGGCTCCCGCCTCCCGCGGCCTA
- Phenotype
-
MIM: 609773
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EID2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3746086] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EID2
- Gene Name
- EP300 interacting inhibitor of differentiation 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153232.3 |
224 |
Silent Mutation |
GCC,GCT |
A35A |
NP_694964.3 |
- Gene
- EID2B
- Gene Name
- EP300 interacting inhibitor of differentiation 2B
There are no transcripts associated with this gene.
View Full Product Details