Product Details

SNP ID
rs199700932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4362349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACGCATGCTCCGGCTAGGGGTC[C/T]GGATGGGCTTGTCGGAAGATCGGAA
Phenotype
MIM: 601768
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPND PubMed Links

Gene Details

Gene
MPND
Gene Name
MPN domain containing
There are no transcripts associated with this gene.

Gene
SH3GL1
Gene Name
SH3 domain containing GRB2 like 1, endophilin A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199943.1 Intron NP_001186872.1
NM_001199944.1 Intron NP_001186873.1
NM_003025.3 Intron NP_003016.1
XM_011528183.2 Intron XP_011526485.1
XM_017027146.1 Intron XP_016882635.1

View Full Product Details