Product Details

SNP ID
rs201335739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38879193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTCTCTGTTGTCCTGGCCTCGT[C/T]CTTGGCAGGTGGCGGGGACTTCTTG
Phenotype
MIM: 604480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RINL PubMed Links

Gene Details

Gene
RINL
Gene Name
Ras and Rab interactor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195833.1 1319 Intron NP_001182762.1
NM_198445.3 1319 Intron NP_940847.1
XM_006723015.2 1319 Intron XP_006723078.1
XM_006723016.2 1319 Intron XP_006723079.1
XM_011526454.2 1319 Intron XP_011524756.1
XM_011526455.2 1319 Intron XP_011524757.1
Gene
SIRT2
Gene Name
sirtuin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193286.1 1319 UTR 3 NP_001180215.1
NM_012237.3 1319 Missense Mutation AAC,GAC N378D NP_036369.2
NM_030593.2 1319 Missense Mutation AAC,GAC N341D NP_085096.1
XM_006723111.1 1319 Missense Mutation AAC,GAC N341D XP_006723174.1
XM_011526654.1 1319 Missense Mutation AAC,GAC N341D XP_011524956.1
XM_011526655.1 1319 Missense Mutation AAC,GAC N308D XP_011524957.1
XM_017026500.1 1319 Missense Mutation AAC,GAC N255D XP_016881989.1

View Full Product Details