Product Details

SNP ID
rs201345208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44287943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGGCTCTGGAAACTTGAGGCTG[A/G]ACTAAAACCCTTACCACACTCTTCA
Phenotype
MIM: 604749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF235 PubMed Links

Gene Details

Gene
ZNF235
Gene Name
zinc finger protein 235
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004234.4 5651 Missense Mutation CCA,TCA P498S NP_004225.3
XM_005259412.4 5651 Missense Mutation CCA,TCA P494S XP_005259469.1

View Full Product Details