Product Details

SNP ID
rs201687831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38879288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATCTATGCTGGCGTGCTCCCTC[C/T]GGACAAGGTCCTCCAGCTCCTTCTG
Phenotype
MIM: 604480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RINL PubMed Links

Gene Details

Gene
RINL
Gene Name
Ras and Rab interactor like
There are no transcripts associated with this gene.

Gene
SIRT2
Gene Name
sirtuin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193286.1 1224 UTR 3 NP_001180215.1
NM_012237.3 1224 Missense Mutation NP_036369.2
NM_030593.2 1224 Missense Mutation NP_085096.1
XM_006723111.1 1224 Missense Mutation XP_006723174.1
XM_011526654.1 1224 Missense Mutation XP_011524956.1
XM_011526655.1 1224 Missense Mutation XP_011524957.1
XM_017026500.1 1224 Missense Mutation XP_016881989.1

View Full Product Details