Product Details

SNP ID
rs201721114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8849017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTATTCTCCTTCCTTCTTCCGC[C/T]GGCGGGTGGTCACCTATGAGGGTGA
Phenotype
MIM: 607963 MIM: 606154
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBD3L1 PubMed Links

Gene Details

Gene
MBD3L1
Gene Name
methyl-CpG binding domain protein 3 like 1
There are no transcripts associated with this gene.

Gene
MUC16
Gene Name
mucin 16, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024690.2 33510 Missense Mutation NP_078966.2
XM_017027486.1 33510 Missense Mutation XP_016882975.1
XM_017027487.1 33510 Missense Mutation XP_016882976.1
XM_017027488.1 33510 Missense Mutation XP_016882977.1
XM_017027489.1 33510 Missense Mutation XP_016882978.1
XM_017027490.1 33510 Missense Mutation XP_016882979.1
XM_017027491.1 33510 Missense Mutation XP_016882980.1
XM_017027492.1 33510 Missense Mutation XP_016882981.1
XM_017027493.1 33510 Missense Mutation XP_016882982.1
XM_017027494.1 33510 Missense Mutation XP_016882983.1
XM_017027495.1 33510 Missense Mutation XP_016882984.1
XM_017027496.1 33510 Missense Mutation XP_016882985.1
XM_017027497.1 33510 Missense Mutation XP_016882986.1
XM_017027498.1 33510 Intron XP_016882987.1
XM_017027499.1 33510 Missense Mutation XP_016882988.1
XM_017027500.1 33510 Missense Mutation XP_016882989.1
XM_017027501.1 33510 Missense Mutation XP_016882990.1

View Full Product Details