Product Details

SNP ID
rs202179952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36340638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCTTCCAACATTCCATACATTC[A/G]TAGGGTTTCTCACGAGTATGTATTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00665 PubMed Links

Gene Details

Gene
LINC00665
Gene Name
long intergenic non-protein coding RNA 665
There are no transcripts associated with this gene.

Gene
ZFP14
Gene Name
ZFP14 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297619.1 1311 Silent Mutation TAC,TAT Y397Y NP_001284548.1
NM_020917.2 1311 Silent Mutation TAC,TAT Y396Y NP_065968.1
XM_017027038.1 1311 Silent Mutation TAC,TAT Y396Y XP_016882527.1

View Full Product Details