Product Details

SNP ID
rs201000917
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:153631757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGAGCTAGACGAGAATGGAGA[C/T]GGGGAGGTGGACTTCCAGGAGTATG
Phenotype
MIM: 614206 MIM: 176940 MIM: 601989
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHTOP PubMed Links

Gene Details

Gene
CHTOP
Gene Name
chromatin target of PRMT1
There are no transcripts associated with this gene.

Gene
S100A1
Gene Name
S100 calcium binding protein A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006271.1 314 Silent Mutation GAC,GAT D67D NP_006262.1
Gene
S100A13
Gene Name
S100 calcium binding protein A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024210.1 314 Intron NP_001019381.1
NM_001024211.1 314 Intron NP_001019382.1
NM_001024212.1 314 Intron NP_001019383.1
NM_001024213.1 314 Intron NP_001019384.1
NM_005979.2 314 Intron NP_005970.1
XM_005245434.3 314 Intron XP_005245491.1
XM_011509862.2 314 Intron XP_011508164.1
XM_011509863.2 314 Intron XP_011508165.1
XM_011509864.1 314 Intron XP_011508166.1
XM_017002034.1 314 Intron XP_016857523.1
XM_017002035.1 314 Intron XP_016857524.1
XM_017002036.1 314 Intron XP_016857525.1

View Full Product Details