Product Details

SNP ID
rs201189703
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:247841403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATTGCGCCCACAGAAGTCCAAC[C/G]TGGAAATCATCAGGGAAGGCAGAAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR11L1 PubMed Links

Gene Details

Gene
OR11L1
Gene Name
olfactory receptor family 11 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001959.1 494 Missense Mutation ACG,AGG T165R NP_001001959.1

View Full Product Details