Product Details

SNP ID
rs199519515
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:24120898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTGAGGTCTCAGGGGCTGGGCT[C/T]TTGGCTTCATCCTTGGGACACACAA
Phenotype
MIM: 605457 MIM: 616832
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL22RA1 PubMed Links

Gene Details

Gene
IL22RA1
Gene Name
interleukin 22 receptor subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021258.3 1835 Silent Mutation AAA,AAG K544K NP_067081.2
XM_011541882.1 1835 Silent Mutation AAA,AAG K408K XP_011540184.1
Gene
MYOM3
Gene Name
myomesin 3
There are no transcripts associated with this gene.

View Full Product Details