Product Details

SNP ID
rs199812597
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:152884436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTCAGAAGATGTGTGACCAGA[A/C]AAAACACAGTAAATGCTGCCCAGCA
Phenotype
MIM: 601148
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMCP PubMed Links

Gene Details

Gene
SMCP
Gene Name
sperm mitochondria associated cysteine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030663.2 159 Missense Mutation AAA,ACA K5T NP_109588.2

View Full Product Details