Product Details

SNP ID
rs199986518
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:203128922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCCCTGGTCTCTGTGCCCGGGAA[C/T]GTGCTGGTGATCTGGGCGGTGAAGG
Phenotype
MIM: 102775
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000674.2 9 Silent Mutation AAC,AAT N27N NP_000665.1
NM_001048230.1 9 Silent Mutation AAC,AAT N27N NP_001041695.1
XM_005244901.1 9 UTR 5 XP_005244958.1
XM_005244902.4 9 UTR 5 XP_005244959.1
XM_017000342.1 9 UTR 5 XP_016855831.1

View Full Product Details