Product Details

SNP ID
rs200583556
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:151807599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGCAGCCTTTCCACATGCTGG[A/C]TACACAGGCTCCGAAGCTTCCCCTT
Phenotype
MIM: 609794 MIM: 602943
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINGO4 PubMed Links

Gene Details

Gene
LINGO4
Gene Name
leucine rich repeat and Ig domain containing 4
There are no transcripts associated with this gene.

Gene
RORC
Gene Name
RAR related orphan receptor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001523.1 2321 Missense Mutation AGC,ATC S456I NP_001001523.1
NM_005060.3 2321 Missense Mutation AGC,ATC S477I NP_005051.2
XM_006711484.3 2321 Missense Mutation AGC,ATC S610I XP_006711547.2

View Full Product Details