Product Details

SNP ID
rs200482554
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:151807499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGACAGCCCCACAGGTGACTC[A/G]GTTTCAGTGCTGAAGAGCTCCTTGT
Phenotype
MIM: 609794 MIM: 602943
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINGO4 PubMed Links

Gene Details

Gene
LINGO4
Gene Name
leucine rich repeat and Ig domain containing 4
There are no transcripts associated with this gene.

Gene
RORC
Gene Name
RAR related orphan receptor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001523.1 2421 Silent Mutation ACC,ACT T489T NP_001001523.1
NM_005060.3 2421 Silent Mutation ACC,ACT T510T NP_005051.2
XM_006711484.3 2421 Silent Mutation ACC,ACT T643T XP_006711547.2

View Full Product Details