Product Details

SNP ID
rs202221385
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:155235732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGTTTGTAAAACGTGTCCTTG[G/T]TGATGTCTACAATGATGGGACTGTC
Phenotype
MIM: 606463
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GBA PubMed Links

Gene Details

Gene
GBA
Gene Name
glucosylceramidase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000157.3 1762 Missense Mutation AAC,ACC N446T NP_000148.2
NM_001005741.2 1762 Missense Mutation AAC,ACC N446T NP_001005741.1
NM_001005742.2 1762 Missense Mutation AAC,ACC N446T NP_001005742.1
NM_001171811.1 1762 Missense Mutation AAC,ACC N359T NP_001165282.1
NM_001171812.1 1762 Missense Mutation AAC,ACC N397T NP_001165283.1
Gene
GBAP1
Gene Name
glucosylceramidase beta pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details