Product Details

SNP ID
rs202168363
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:166849036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCACTTTGACAGCTTTGGCCTC[C/T]GTCTGCCTCGGGATATCACAGAGCT
Phenotype
MIM: 612763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POGK PubMed Links

Gene Details

Gene
POGK
Gene Name
pogo transposable element with KRAB domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314014.1 937 Missense Mutation CGT,TGT R35C NP_001300943.1
NM_017542.4 937 Missense Mutation CGT,TGT R153C NP_060012.3
XM_005245370.3 937 Missense Mutation CGT,TGT R153C XP_005245427.1
XM_011509814.2 937 Missense Mutation CGT,TGT R153C XP_011508116.1
XM_011509815.1 937 Missense Mutation CGT,TGT R153C XP_011508117.1
XM_017001914.1 937 Missense Mutation CGT,TGT R153C XP_016857403.1
XM_017001915.1 937 Missense Mutation CGT,TGT R35C XP_016857404.1
Gene
TADA1
Gene Name
transcriptional adaptor 1
There are no transcripts associated with this gene.

View Full Product Details