Product Details

SNP ID
rs201932234
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:182904355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGATAGATGCATTAGTTTCACA[C/T]TGTCAGCTTTGGACACCACAAAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHCBP1L PubMed Links

Gene Details

Gene
SHCBP1L
Gene Name
SHC binding and spindle associated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030933.2 1697 Missense Mutation AAT,AGT N471S NP_112195.2

View Full Product Details