Product Details

SNP ID
rs201908321
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:26320243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTCAGCATCCAGCAACATAAGC[A/G]GAGGCATTTTCCTTTTCTTCGTGGC
Phenotype
MIM: 114280 MIM: 609151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AIM1L PubMed Links

Gene Details

Gene
AIM1L
Gene Name
absent in melanoma 1-like
There are no transcripts associated with this gene.

Gene
CD52
Gene Name
CD52 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001803.2 225 Missense Mutation AGA,GGA R43G NP_001794.2
Gene
UBXN11
Gene Name
UBX domain protein 11
There are no transcripts associated with this gene.

View Full Product Details