Product Details

SNP ID
rs199716028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5115310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATCCTTCTTCATCCTCGATCTT[A/G]GGAGCCAAGTAGTATTTTAAGTGTC
Phenotype
MIM: 176740 MIM: 617019
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCNA PubMed Links

Gene Details

Gene
PCNA
Gene Name
proliferating cell nuclear antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002592.2 998 Silent Mutation CCC,CCT P253P NP_002583.1
NM_182649.1 998 Intron NP_872590.1
Gene
PCNA-AS1
Gene Name
PCNA antisense RNA 1
There are no transcripts associated with this gene.

Gene
TMEM230
Gene Name
transmembrane protein 230
There are no transcripts associated with this gene.

View Full Product Details