Product Details
- SNP ID
-
rs200752417
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31450069 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGGAATCTTTATGGCAAATGCCG[A/T]TACAGATGCTCCAAGAAGGAAAGAG
- Phenotype
-
MIM: 616076
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB123
PubMed Links
Gene Details
- Gene
- DEFB123
- Gene Name
- defensin beta 123
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153324.3 |
279 |
Silent Mutation |
CGA,CGT |
R33R |
NP_697019.1 |
View Full Product Details