Product Details

SNP ID
rs200710399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31820355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGGAGGGCAGTGCTGGGCCCCC[A/G]GCAGCCCTGCCCCAGCAGACTGCGA
Phenotype
MIM: 606566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYLK2 PubMed Links

Gene Details

Gene
MYLK2
Gene Name
myosin light chain kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033118.3 384 Silent Mutation CCA,CCG P94P NP_149109.1

View Full Product Details