Product Details

SNP ID
rs201713999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30337089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCACCAGTAACACAGCAAGTTG[G/T]TTCAACTCCTGGAGAAGATTCCAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP27-1 PubMed Links

Gene Details

Gene
KRTAP27-1
Gene Name
keratin associated protein 27-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077711.1 606 Missense Mutation NP_001071179.1

View Full Product Details