Product Details

SNP ID
rs200504770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41367598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCCTTGGCCAGAGAAGAGAAC[A/G]GGTAGGAGCACAGGTGACCTGCATT
Phenotype
MIM: 188595
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100996598 PubMed Links

Gene Details

Gene
LOC100996598
Gene Name
uncharacterized LOC100996598
There are no transcripts associated with this gene.

Gene
TEF
Gene Name
TEF, PAR bZIP transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145398.2 266 Silent Mutation ACA,ACG T22T NP_001138870.1
NM_003216.3 266 Intron NP_003207.1
Gene
ZC3H7B
Gene Name
zinc finger CCCH-type containing 7B
There are no transcripts associated with this gene.

View Full Product Details